Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153501.2 | NP_705721.2 | 4 | 1027 | ||
GenBank mRNA | AK016755.1 | - | 6 | 994 | ||
AK082821.1 | - | 4 | 1033 | |||
AK154080.1 | - | 4 | 991 | |||
AK159175.1 | - | 4 | 1019 | |||
AK167861.1 | - | 4 | 735 | |||
BC106184.1 | - | 4 | 981 | 30241166 |