Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamstr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172418.2 | NP_766006.2 | 8 | 855 | ||
GenBank mRNA | AK017382.1 | - | 7 | 1163 | ||
AK036109.1 | - | 5 | 973 | |||
AK052976.1 | - | 2 | 2008 | |||
BC107175.1 | - | 3 | 374 | |||
BC107176.1 | - | 3 | 376 | |||
BC119481.1 | - | 3 | 374 | |||
BC119482.1 | - | 3 | 374 | |||
BC127227.1 | - | 3 | 376 | |||
BC127228.1 | - | 3 | 376 | |||
DQ534901.1 | - | 8 | 839 |