Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278554.1 | NP_001265483.1 | 5 | 870 | ||
NM_001278555.1 | NP_001265484.1 | 5 | 678 | |||
NM_006357.3 | NP_006348.1 | 5 | 909 | |||
NM_182678.2 | NP_872619.1 | 6 | 710 | |||
XM_005246244.2 | XP_005246301.1 | 5 | 866 | |||
GenBank mRNA | AB017644.1 | - | 5 | 552 | ||
AF085362.1 | - | 5 | 610 | |||
AK314145.1 | - | 5 | 880 | |||
BC003554.1 | - | 5 | 592 | 3537280 | ||
BC092407.1 | - | 5 | 849 | 6180242 | ||
BQ932163.1 | - | 6 | 710 | |||
BT019345.1 | - | 4 | 491 |
Gene Symbol: UBE2E4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110506.1 | 1 | 494 | |||
GenBank mRNA | AF136176.1 | - | 1 | 494 |