Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173182.2 | NP_775274.2 | 26 | 5957 | ||
GenBank mRNA | AB098596.1 | - | 26 | 5893 | ||
AK039335.1 | - | 1 | 631 | |||
AK050037.1 | - | 1 | 804 | |||
AK147346.1 | - | 27 | 5968 | |||
AK220297.1 | - | 26 | 5957 | |||
DQ192037.1 | - | 25 | 5864 |