Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175224.4 | NP_780433.1 | 8 | 749 | ||
GenBank mRNA | AK006484.1 | - | 3 | 341 | ||
AK077694.1 | - | 8 | 750 | |||
AK129054.1 | - | 3 | 2856 | |||
AK152138.1 | - | 8 | 768 | |||
AK153231.1 | - | 8 | 768 | |||
AK153944.1 | - | 8 | 757 | |||
BC027110.1 | - | 3 | 289 | |||
BC096469.1 | - | 8 | 740 | 3603051 |