Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122666.2 | NP_001116138.1 | 13 | 1414 | ||
NM_178622.5 | NP_848737.3 | 12 | 1503 | |||
GenBank mRNA | AF469109.1 | - | 12 | 1443 | ||
AK010274.1 | - | 12 | 1407 | |||
AK043652.1 | - | 12 | 1446 | |||
AK045475.1 | - | 12 | 1457 | |||
AK138947.1 | - | 12 | 1454 | |||
AK147512.1 | - | 3 | 8865 | |||
AK150633.1 | - | 12 | 1441 | |||
BC036562.1 | - | 7 | 876 | |||
BC040088.1 | - | 10 | 1116 | |||
BC055764.1 | - | 12 | 1456 | 6402735 | ||
BC062176.1 | - | 13 | 1281 | 6514171 |