Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012978.1 | - | 1, 2 | 227 | ||
BC052657.1 | - | 1, 2 | 219 | 30042589 | ||
BY776788.1 | - | 1, 2 | 233 |