Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161415.1 | NP_001154887.1 | 4 | 731 | ||
NM_001161416.1 | NP_001154888.1 | 3 | 666 | |||
NM_001161417.1 | NP_001154889.1 | 2 | 437 | |||
NM_005291.2 | NP_005282.1 | 3 | 502 | |||
XM_017003833.1 | XP_016859322.1 | 2 | 858 | |||
GenBank mRNA | AK126849.1 | - | 1 | 1692 | ||
AK292725.1 | - | 2 | 307 | |||
AK313949.1 | - | 2 | 193 | |||
BC031653.1 | - | 2 | 116 | 5174146 | ||
BC039595.1 | - | 2 | 121 | |||
DA154324.1 | - | 2 | 437 | |||
DA314295.1 | - | 3 | 214 | |||
DA366480.1 | - | 3 | 538 | |||
DA390892.1 | - | 2 | 416 | |||
JF810891.1 | - | 1 | 36 | |||
Y12546.1 | - | 3 | 194 | |||
Z94154.1 | - | 2 | 130 |