Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182994.2 | NP_892039.1 | 6 | 1690 | ||
GenBank mRNA | AK147525.1 | - | 6 | 1690 | ||
BC048170.1 | - | 6 | 1690 | 6821976 | ||
BC059931.1 | - | 6 | 1690 |