Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023343.2 | NP_075832.1 | 6, 7 | 618 | ||
GenBank mRNA | AF117831.1 | - | 1, 2 | 72 | ||
AK003007.1 | - | 5, 6 | 1591 | |||
AK005408.1 | - | 6, 7 | 641 | |||
BC011427.1 | - | 6, 7 | 588 | |||
BC026953.1 | - | 6, 7 | 618 | 5133980 | ||
BC027439.1 | - | 6, 7 | 507 | 4164301 |