Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023395.2 | NP_075884.1 | 3 | 556 | ||
GenBank mRNA | AK009971.1 | - | 3 | 559 | ||
AK018575.1 | - | 3 | 417 | |||
AK132541.1 | - | 3 | 557 | |||
AK142707.1 | - | 3 | 425 | |||
AK161606.1 | - | 3 | 422 | |||
BC025850.1 | - | 3 | 372 | |||
BC038122.1 | - | 3 | 398 | |||
BC132073.1 | - | 3 | 485 | 40130350 | ||
BC132469.1 | - | 3 | 485 | 40130746 | ||
BC145305.1 | - | 3 | 485 | 9053859 |