Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023503.3 | NP_075992.2 | 2 | 1654 | ||
GenBank mRNA | AB433625.1 | - | 2 | 1157 | ||
AK048800.1 | - | 2 | 1650 | |||
AK083144.1 | - | 2 | 1653 | |||
BC138229.1 | - | 2 | 1385 | 8861248 |