Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023873.3 | NP_076362.2 | 13 | 1269 | ||
GenBank mRNA | AF242319.1 | - | 13 | 1305 | ||
AK133048.1 | - | 13 | 1275 | |||
BC050819.1 | - | 13 | 1261 | 6517145 | ||
BC071260.1 | - | 13 | 1269 | 6336351 |