Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026062.4 | NP_080338.1 | 5 | 581 | ||
GenBank mRNA | AK008504.1 | - | 5 | 525 | ||
AK013580.1 | - | 5 | 545 | |||
AK038811.1 | - | 5 | 539 | |||
AK048979.1 | - | 5 | 538 | |||
AK050305.1 | - | 5 | 538 | |||
AK053273.1 | - | 4 | 386 | |||
AK153488.1 | - | 5 | 536 | |||
BC055842.1 | - | 5 | 513 | |||
BC061013.1 | - | 5 | 533 | |||
BC095935.1 | - | 5 | 515 | |||
BC116683.1 | - | 5 | 526 | 40106565 |