Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026070.3 | NP_080346.1 | 4, 5 | 468 | ||
NM_027487.1 | NP_081763.1 | 4, 5 | 468 | |||
GenBank mRNA | AK004206.1 | - | 4, 5 | 383 | ||
AK009358.1 | - | 4, 5 | 364 | |||
AK013835.1 | - | 4, 5 | 473 | |||
AK017732.1 | - | 4, 5 | 396 | |||
AK158822.1 | - | 4, 5 | 469 | |||
BC029256.1 | - | 4, 5 | 365 | 3661472 |