Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nanp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026086.2 | NP_080362.1 | 2 | 868 | ||
GenBank mRNA | AK005566.1 | - | 2 | 844 | ||
AK021344.1 | - | 2 | 855 | |||
BC018527.1 | - | 2 | 839 | 3596532 | ||
BC083086.1 | - | 2 | 868 | 6515732 |