Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 16, 17 | 2512 | ||
GenBank mRNA | AK007365.1 | - | 10, 11 | 1217 | ||
AK011477.1 | - | 6, 7 | 596 | |||
AY033907.1 | - | 16, 17 | 2297 | |||
AY033908.1 | - | 16, 17 | 2314 | |||
BC014838.1 | - | 9, 10 | 997 | 3588322 | ||
BC027145.1 | - | 8, 9 | 932 |