Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026111.3 | NP_080387.2 | 6 | 952 | ||
GenBank mRNA | AK007555.1 | - | 6 | 997 | ||
AK007701.1 | - | 6 | 980 | |||
AK035587.1 | - | 6 | 952 | |||
AK045687.1 | - | 6 | 956 | |||
AK136746.1 | - | 95 | ||||
BC058181.1 | - | 6 | 975 | 5325516 |