Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166397.1 | NP_001159869.1 | 6 | 3409 | ||
NM_001166398.1 | NP_001159870.1 | 6 | 3352 | |||
NM_026139.4 | NP_080415.3 | 5 | 3516 | |||
GenBank mRNA | AK014329.1 | - | 6 | 3185 | ||
AK017587.1 | - | 1 | 537 | |||
AK034185.1 | - | 6 | 3185 | |||
AK034621.1 | - | 6 | 3183 | |||
AK043440.1 | - | 6 | 3186 | |||
AK044926.1 | - | 6 | 3183 | |||
AK053354.1 | - | 5 | 3290 | |||
AK077990.1 | - | 1 | 2294 | |||
AK081299.1 | - | 6 | 3128 | |||
AK172960.1 | - | 6 | 3409 | |||
BC006943.1 | - | 1 | 1141 | |||
BC049088.1 | - | 6 | 3164 | |||
BC052383.1 | - | 6 | 3158 | |||
BC054839.1 | - | 3 | 2748 | 6490133 |