Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aasdhppt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026276.3 | NP_080552.3 | 6 | 1220 | ||
GenBank mRNA | AK010557.1 | - | 6 | 1180 | ||
AK014188.1 | - | 6 | 1163 | |||
AK018191.1 | - | 5 | 866 | |||
AK028700.1 | - | 6 | 1178 | |||
AK029523.1 | - | 6 | 1162 | |||
AK031766.1 | - | 5 | 1042 | |||
AK076099.1 | - | 6 | 1175 | |||
AK076866.1 | - | 6 | 1194 | |||
AK081818.1 | - | 6 | 1197 | |||
AK088977.1 | - | 5 | 1038 | |||
AK144671.1 | - | 6 | 1194 | |||
BC030043.1 | - | 6 | 1155 | 1348651 | ||
BC038013.1 | - | 6 | 1191 | 3497754 |