Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026313.1 | NP_080589.1 | 5 | 521 | ||
GenBank mRNA | AK014362.1 | - | 5 | 514 | ||
AK019464.1 | - | 5 | 574 | |||
AK078090.1 | - | 5 | 568 | |||
AK151228.1 | - | 5 | 569 | |||
AK151839.1 | - | 5 | 569 | |||
BC009092.1 | - | 5 | 521 | 3157532 |