Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026360.3 | NP_080636.2 | 5 | 537 | ||
GenBank mRNA | AK010310.1 | - | 5 | 530 | ||
AK076982.1 | - | 5 | 522 | |||
AK169595.1 | - | 3 | 160 | |||
BC060168.1 | - | 5 | 517 | |||
BC095944.1 | - | 5 | 514 | 4935970 | ||
BC132242.1 | - | 5 | 529 | 40130519 | ||
BC132244.1 | - | 5 | 529 | 40130521 |