Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026384.3 | NP_080660.1 | 8 | 1806 | ||
GenBank mRNA | AK002443.1 | - | 8 | 1812 | ||
AK003803.1 | - | 1 | 245 | |||
BC043447.1 | - | 8 | 1806 | 5125951 |