Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026406.3 | NP_080682.3 | 4, 5 | 627 | ||
GenBank mRNA | AK008329.1 | - | 4, 5 | 481 | ||
AK011584.1 | - | 4, 5 | 557 | |||
AK042216.1 | - | 5, 6 | 622 | |||
AK131966.1 | - | 4, 5 | 547 | |||
AK159772.1 | - | 4, 5 | 520 | |||
BC023113.1 | - | 4, 5 | 574 | 4015975 |