Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199151.1 | NP_001186080.1 | 4 | 718 | ||
NM_001199153.1 | NP_001186082.1 | 4 | 623 | |||
NM_001199154.1 | NP_001186083.1 | 4 | 893 | |||
NM_026408.4 | NP_080684.3 | 4 | 718 | |||
GenBank mRNA | AF348447.1 | - | 4 | 646 | ||
AK013040.1 | - | 2 | 451 | |||
AK016941.1 | - | 4 | 635 | |||
AK044974.1 | - | 4 | 916 | |||
AK081942.1 | - | 4 | 897 | |||
AK082079.1 | - | 4 | 722 | |||
AK132346.1 | - | 4 | 821 | |||
AK160111.1 | - | 4 | 722 | |||
BC098489.1 | - | 4 | 622 | 30096466 | ||
BC130225.1 | - | 4 | 608 | |||
BC152544.1 | - | 4 | 606 |