Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saraf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026432.3 | NP_080708.3 | 6 | 1593 | ||
GenBank mRNA | AK007787.1 | - | 6 | 1520 | ||
AK075744.1 | - | 6 | 1484 | |||
AK077118.1 | - | 6 | 1580 | |||
AK088728.1 | - | 5 | 2072 | |||
AK093942.1 | - | 6 | 1571 | |||
AK163218.1 | - | 7 | 1505 | |||
BC013497.1 | - | 6 | 1534 | 4222385 | ||
BC022616.1 | - | 6 | 1571 | 4187981 | ||
BC024888.1 | - | 6 | 1529 | 3988822 |