Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apopt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026511.2 | NP_080787.1 | 5 | 524 | ||
GenBank mRNA | AK002873.1 | - | 5 | 526 | ||
AK005403.1 | - | 5 | 512 | |||
AK010493.1 | - | 5 | 514 | |||
AK012648.1 | - | 5 | 522 | |||
AK029408.1 | - | 5 | 500 | |||
BC071262.1 | - | 5 | 524 | 6509633 |