Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026530.5 | NP_080806.4 | 11 | 1284 | ||
GenBank mRNA | AK012128.1 | - | 10 | 1239 | ||
AK146973.1 | - | 11 | 1269 | |||
AK160356.1 | - | 11 | 1253 | |||
AK172381.1 | - | 10 | 1027 | |||
BC017625.1 | - | 10 | 1233 |