Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026530.5 | NP_080806.4 | 13 | 1551 | ||
GenBank mRNA | AK012128.1 | - | 12 | 1506 | ||
AK146973.1 | - | 13 | 1536 | |||
AK160356.1 | - | 13 | 1521 | |||
AK172381.1 | - | 12 | 1294 | |||
BC017625.1 | - | 12 | 1500 |