Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026533.3 | NP_080809.1 | 5 | 418 | ||
GenBank mRNA | AK012479.1 | - | 5 | 363 | ||
AK018703.1 | - | 5 | 362 | |||
AK076060.1 | - | 5 | 362 | |||
AK088237.1 | - | 4 | 995 | |||
AK151043.1 | - | 5 | 363 | |||
AK151671.1 | - | 5 | 364 | |||
AK161798.1 | - | 3 | 1410 | |||
AK166691.1 | - | 5 | 363 | |||
AV498110.1 | - | 4 | 325 | |||
BC011192.1 | - | 5 | 361 | 4016679 | ||
BC085269.1 | - | 2 | 1516 | |||
BC090397.1 | - | 5 | 345 | 5150054 | ||
BC106137.1 | - | 5 | 346 | 4990874 | ||
BC125602.1 | - | 5 | 362 | 40130117 | ||
BC125604.1 | - | 5 | 362 | 40130119 |