Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam92a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026558.4 | NP_080834.3 | 2 | 459 | ||
GenBank mRNA | AK013129.1 | - | 2 | 290 | ||
AK019574.1 | - | 2 | 287 | |||
AK032690.1 | - | 2 | 440 | |||
AK032976.1 | - | 1 | 756 | |||
AK078438.1 | - | 2 | 440 | |||
AK088705.1 | - | 2 | 440 | |||
BC020162.1 | - | 253 | 3991990 | |||
BC055848.1 | - | 2 | 269 |