Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026615.4 | NP_080891.1 | 5 | 2126 | ||
GenBank mRNA | AK049849.1 | - | 5 | 2127 | ||
AK165008.1 | - | 2 | 3126 |