Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700067P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026625.2 | NP_080901.2 | 3 | 632 | ||
GenBank mRNA | AK018887.1 | - | 3 | 621 | ||
BC048440.1 | - | 3 | 622 | 6741836 |