Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtsf1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026630.2 | NP_080906.1 | 2 | 651 | ||
GenBank mRNA | AK018886.1 | - | 1 | 591 | ||
AK019131.1 | - | 2 | 652 | |||
AK160150.1 | - | 1 | 551 | |||
BC048439.1 | - | 1 | 524 | 6774366 |