Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026880.2 | NP_081156.2 | 3 | 771 | ||
GenBank mRNA | AB053476.1 | - | 3 | 703 | ||
AF316872.1 | - | 3 | 749 | |||
AK050322.1 | - | 3 | 751 | |||
AK165480.1 | - | 3 | 750 | |||
AK188455.1 | - | 2 | 299 | |||
AK213702.1 | - | 2 | 112 | |||
AK220318.1 | - | 3 | 725 | |||
BC018561.1 | - | 3 | 597 | |||
BC018594.1 | - | 3 | 734 | |||
BC067066.1 | - | 3 | 739 | 30542123 | ||
BC069968.1 | - | 4 | 629 |