Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026949.3 | NP_081225.1 | 2, 3 | 262 | ||
GenBank mRNA | AK007581.1 | - | 2, 3 | 326 | ||
AK075727.1 | - | 2, 3 | 232 | |||
AK150757.1 | - | 2, 3 | 343 | |||
AK153907.1 | - | 2, 3 | 265 | |||
AY515608.1 | - | 1, 2 | 229 | |||
BC004040.1 | - | 2, 3 | 262 | 3496188 |