Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026987.2 | NP_081263.2 | 5 | 862 | ||
GenBank mRNA | AK122314.2 | - | 5 | 825 | ||
BC009147.1 | - | 5 | 821 | 2647184 | ||
BC046781.1 | - | 5 | 815 | 5716182 | ||
BC138204.1 | - | 5 | 801 | 8861223 | ||
BC138205.1 | - | 5 | 801 | 8861224 |