Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026993.3 | NP_081269.1 | 3 | 623 | ||
GenBank mRNA | AK010430.1 | - | 3 | 589 | ||
AK128909.1 | - | 3 | 178 | |||
AK143507.1 | - | 3 | 640 | |||
AK148907.1 | - | 3 | 624 | |||
AK149185.1 | - | 3 | 275 | |||
BC034505.1 | - | 3 | 574 | 5134831 |