Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027000.4 | NP_081276.2 | 2 | 106 | ||
GenBank mRNA | AF325354.1 | - | 2 | 90 | ||
AF348208.1 | - | 2 | 113 | |||
AK011580.1 | - | 2 | 117 | |||
AK141852.1 | - | 2 | 116 | |||
AK164330.1 | - | 2 | 107 | |||
AK166711.1 | - | 2 | 112 | |||
BC005514.1 | - | 2 | 87 | |||
BC025431.1 | - | 2 | 111 | 5007174 |