Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027102.3 | NP_081378.1 | 5 | 788 | ||
GenBank mRNA | AF361882.1 | - | 5 | 773 | ||
AK009223.1 | - | 5 | 763 | |||
AK156399.1 | - | 5 | 786 | |||
BC019460.1 | - | 5 | 772 | 4188050 | ||
BC049249.1 | - | 5 | 768 | 5039892 | ||
BG077536.2 | - | 3 | 222 |