Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027193.2 | NP_081469.2 | 8 | 1097 | ||
GenBank mRNA | AK010475.1 | - | 8 | 1087 | ||
BC060372.1 | - | 8 | 1097 | 6391546 |