Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 3 | 561 | ||
GenBank mRNA | AK010545.1 | - | 3 | 493 | ||
AK045768.1 | - | 3 | 520 | |||
AK083654.1 | - | 3 | 482 | |||
AK085975.1 | - | 3 | 513 | |||
AK086015.1 | - | 3 | 497 | |||
AK086059.1 | - | 3 | 516 | |||
AK088246.1 | - | 3 | 1026 | |||
AK149407.1 | - | 3 | 490 | |||
AK160039.1 | - | 3 | 498 | |||
AK166012.1 | - | 3 | 514 | |||
AK206565.1 | - | 2 | 140 | |||
BC026789.1 | - | 3 | 488 | 4188168 |