Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 423 | ||
GenBank mRNA | BC108336.1 | - | 4 | 401 | 30462068 |
Gene Symbol: Gm13139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083918.1 | NP_001077387.1 | 7 | 560 | ||
GenBank mRNA | BC024323.1 | - | 7 | 560 | 4242170 |
Gene Symbol: Gm13152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039209.2 | NP_001034298.1 | 237 | |||
GenBank mRNA | BC058613.1 | - | 237 | 30044792 |
Gene Symbol: Gm13154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014397.4 | NP_001014419.2 | 6 | 516 | ||
GenBank mRNA | BC085112.1 | - | 6 | 498 | 30731985 | |
CF896923.1 | - | 4 | 392 |
Gene Symbol: Gm13212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205101.1 | NP_001192030.1 | 3 | 214 |
Gene Symbol: Gm13251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085522.2 | NP_001078991.1 | 4 | 288 | ||
GenBank mRNA | CJ326493.1 | - | 4 | 304 |
Gene Symbol: Zfp534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127188.2 | NP_001120660.1 | 4 | 468 |
Gene Symbol: Znf41-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC096621.1 | - | 343 | 30464103 | ||
CN537761.1 | - | 530 |