Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027292.2 | NP_081568.1 | 5 | 618 | ||
GenBank mRNA | AK011003.1 | - | 5 | 619 | ||
AK162634.1 | - | 2 | 2351 | |||
BC104344.1 | - | 4 | 477 | 40049489 | ||
BC104345.1 | - | 4 | 477 | 40049491 |