Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027315.4 | NP_081591.3 | 5 | 819 | ||
GenBank mRNA | AK009324.1 | - | 4 | 401 | ||
AK146707.1 | - | 5 | 632 | |||
AK177704.1 | - | 4 | 371 | |||
AK214106.1 | - | 4 | 338 | |||
AY112699.1 | - | 5 | 812 | |||
AY945937.1 | - | 5 | 716 | |||
BC030044.1 | - | 2 | 144 | |||
BC060966.1 | - | 3 | 212 | |||
BC082275.1 | - | 5 | 642 | 6310111 |