Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810408A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027419.3 | NP_081695.2 | 9 | 1592 | ||
GenBank mRNA | AK160144.1 | - | 8 | 1351 | ||
AK171674.1 | - | 8 | 1503 | |||
BC005538.1 | - | 8 | 884 | |||
BC051546.1 | - | 8 | 1418 | 5255109 | ||
BC055710.1 | - | 7 | 2146 | |||
BC069874.1 | - | 9 | 1592 | 4216824 |