Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038015.1 | NP_001033104.1 | 3 | 278 | ||
GenBank mRNA | AK014950.1 | - | 3 | 288 | ||
AK015527.1 | - | 3 | 288 | |||
AK016785.1 | - | 333 | ||||
AK017588.1 | - | 3 | 287 | |||
AK051596.1 | - | 3 | 287 | |||
AK141408.1 | - | 3 | 290 | |||
AK166601.1 | - | 3 | 280 | |||
AK168516.1 | - | 3 | 297 | |||
AK171299.1 | - | 3 | 273 | |||
AK220275.1 | - | 3 | 239 | |||
BC004084.1 | - | 3 | 278 | 3593102 |