Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpda2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038015.1 | NP_001033104.1 | 7 | 1007 | ||
GenBank mRNA | AK014950.1 | - | 7 | 1016 | ||
AK015527.1 | - | 7 | 1017 | |||
AK016785.1 | - | 1056 | ||||
AK017588.1 | - | 7 | 1016 | |||
AK141408.1 | - | 7 | 1019 | |||
AK166601.1 | - | 7 | 1009 | |||
AK168516.1 | - | 7 | 1026 | |||
AK171299.1 | - | 7 | 1002 | |||
AK220275.1 | - | 6 | 5593 | |||
BC004084.1 | - | 7 | 1007 | 3593102 |