Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027727.2 | NP_082003.1 | 4 | 509 | ||
GenBank mRNA | AK016957.1 | - | 4 | 438 | ||
AK049909.1 | - | 4 | 474 | |||
AK080149.1 | - | 4 | 486 | |||
AK081242.1 | - | 4 | 501 | |||
AK149628.1 | - | 4 | 483 | |||
AK149879.1 | - | 4 | 484 | |||
AK149891.1 | - | 4 | 483 | |||
AK154794.1 | - | 4 | 498 | |||
BC038276.1 | - | 4 | 511 | 4458792 | ||
BC092283.1 | - | 4 | 495 |