Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006418.4 | NP_006409.3 | 5 | 900 | ||
GenBank mRNA | AF097021.1 | - | 5 | 817 | ||
AK300947.1 | - | 5 | 780 | |||
AY358567.1 | - | 5 | 817 | |||
BC047740.1 | - | 4 | 531 | |||
BC117329.1 | - | 5 | 882 | 40125880 |